Order Entry
United States
ContactUsLinkComponent
Br-BTXI ≥98.0% (by HPLC, total nitrogen)
Br-BTXI ≥98.0% (by HPLC, total nitrogen)
Catalog # TCB6102-200MG
Supplier:  TCI America
New Product
CAS Number:  
Br-BTXI ≥98.0% (by HPLC, total nitrogen)
Catalog # TCB6102-200MG
Supplier:  TCI America
Supplier Number:  B6102-200MG
New Product
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    200 mg
  • Cat. No.:
    TCB6102-200MG

Specifications

Specification Test Results

  • Appearance:
    Light yellow to Brown powder to crystal
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (with Total Nitrogen):
    min. 98.0 %

Specification Test Results

About this item

 :
 :452.37 g/mol
CAS Number:2253748-36-4
Formula:C₂₃H₁₈BrNO₂S