Order Entry
United States
ContactUsLinkComponent
3-Cyanopyridine-N-oxide ≥98.0% (by GC, titration analysis)
Irritant
3-Cyanopyridine-N-oxide ≥98.0% (by GC, titration analysis)
Catalog # TCC3416-1G
Supplier:  TCI America
New Product
CAS Number:  
Irritant
3-Cyanopyridine-N-oxide ≥98.0% (by GC, titration analysis)
Catalog # TCC3416-1G
Supplier:  TCI America
Supplier Number:  C3416-1G
New Product
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Size:
    1 g
  • Cat. No.:
    TCC3416-1G

Specifications

Specification Test Results

  • Appearance:
    White to Light yellow to Light orange powder to crystal
  • Purity (GC):
    min. 98.0 %
  • Purity (Nonaqueous Titration):
    min. 98.0 %
  • Melting point:
    175.0 to 179.0 deg-C

Specification Test Results

About this item

 :
 :120.11 g/mol
EINECS:238-975-1
 :GHS07
CAS Number:14906-64-0
Formula:C₆H₄N₂O
MDL Number:MFCD00129037