Hamster (Armenian) monoclonal IgG2 antibody against mouse T cell receptor gamma/delta, APC-conjugated.
- T cells
- T cells other subsets
The GL3 antibody reacts with the δ chain of the murine T cell receptor γ/δ (TCRγ/δ or TCR gamma/delta), a subtype of the TCR involved in the recognition of both peptide and lipid antigens. TCRγ/δ comprises a heterodimer (~80 kDa in humans) of disulfide-linked γ and δ subunits that associates with CD3 on the cell surface. It is a member of the immunoglobulin superfamily. TCRγ/δ is expressed on a subpopulation of T cells in the circulation but may be found on up to 50% of the T cells in epithelial cell-rich tissues. These cells have roles in oral and tumor-associated tolerance as well as autoimmune disease and have been described as a link between the adaptive and innate immune responses. Once activated, they secrete effector cytokines in a subtype- and context-specific manner. Most γ/δ T cells are CD4-/CD8-, though some express CD8. The GL3 antibody recognizes an epitope in the constant region of the δ chain and can reportedly activate TCRγ/δ+ cells.
Type:
Antigen: TCR Gamma/Delta
Clonality: Monoclonal
Clone: GL3
Conjugation: APC
Epitope:
Host: Hamster
Isotype: IgG2 kappa
Reactivity: Mouse
- Catalog No:
- MSPP-60104AZ1
- MSPP-60104AZ
- Antigen Name:
- TCR Gamma/Delta
- TCR Gamma/Delta
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- APC (Allophycocyanin)
- APC (Allophycocyanin)
- Clone:
- GL3
- GL3
- Reactivity:
- Mouse
- Mouse
- Host:
- Hamster
- Hamster
- Gene ID:
- 110066/110067
- 110066/110067
- Isotype:
- IgG2 kappa
- IgG2 kappa