To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Antigen Name:CD69
- Clonality:Monoclonal
- Clone:H1.2F3
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Hamster
- Isotype:IgG
- Reactivity:Mouse
- Size:100 µg
- Environmentally Preferable:
- Gene ID:12515
- Cat. No.:MSPP-100-1617
Hamster (Armenian) monoclonal antibody against mouse CD69, FITC-conjugated.
- B cells
- Macrophages
- Monocytes
- Nk cells
- T cells
This monoclonal antibody (clone H1.2F3) reacts with mouse cluster of differentiation 69 (CD69), a type II transmembrane glycoprotein. CD69 is a 60 kDa disulfide bonded homodimer expressed on activated T cells, B cells, NK cells, neutrophils, and monocytes, where induction of CD69 rapidly occurs upon activation. CD69 is constitutively expressed on platelets and a subset of thymocytes. It functions as a co-stimulatory molecule that plays a role in activating and promoting the proliferation of T cells. Additionally, it can serve as an indicator for thymocytes engaged in T cell receptor (TCR)-mediated positive selection. Furthermore, the protein may act to transmit signals in natural killer cells and platelets. It has also been documented that co-stimulation with the H1.2F3 antibody clone enhances the activation of T cells and macrophages. CD69 dysfunction is associated with diseases such as coccidioidomycosis and asthma. Use this recombinant monoclonal antibody in applications requiring detection of CD69, such as flow cytometry (FC).
Type:
Antigen: CD69
Clonality: Monoclonal
Clone: H1.2F3
Conjugation: FITC
Epitope:
Host: Hamster
Isotype: IgG
Reactivity: Mouse