To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Antigen Name:TBR1 (IDDAS)
- Clonality:Polyclonal
- Clone:Poly
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Size:100 µl
- Gene ID:10716
- Cat. No.:MSPP-100-1341
Specifications
About this item
Rabbit polyclonal antibody against human, mouse, rat TBR1 (IDDAS).
- Neural cells Psc derived
- Neurons
This rabbit polyclonal antibody reacts with human, mouse and rat T-box brain 1 (TBR1), a transcription factor highly expressed and involved in human development; mutations of the T-box gene have been linked to developmental disorders as well as the manifestation of certain cancers. Use this polyclonal antibody in applications requiring detection of TBR1, such as ELISA, western blot (WB), immunohistochemistry (IHC), immunocytochemistry (ICC), and immunofluorescence (IF). This antibody clone has been verified for labeling TBR1-positive cortical neurons in hPSC-derived cerebral organoids generated with STEMdiff™ Cerebral Organoid Kit (Catalog #08570) and STEMdiff™ Cerebral Organoid Maturation Kit (Catalog #08571). The T-box transcription factor family consists of five subfamilies, with the TBR1 family of proteins responsible for brain development, primarily through the function of T-box proteins that act as transcriptional activators and inhibitors. In particular, the function of TBR1 in brain development is primarily facilitated by the differentiation of neural stem cells (NSCs) in both developing and adult brains. Structurally, T-box transcription factors contain a conserved DNA-binding motif named T-box domain, spanning 180-200 amino acid residues to bind DNA. Studies involving TBR1-deficient mice found defects in the frontal cortex and layer 6 differentiation.
Type:
Antigen: TBR1 (IDDAS)
Clonality: Polyclonal
Clone: Poly
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat