Order Entry
United States
ContactUsLinkComponent
Anti-CD184 (CXCR4) Mouse Monoclonal Antibody [clone: 12G5] (APC)
Anti-CD184 (CXCR4) Mouse Monoclonal Antibody [clone: 12G5] (APC)
Certificates
undefined
Anti-CD184 (CXCR4) Mouse Monoclonal Antibody [clone: 12G5] (APC)

Specifications

  • Catalog No:
  • MSPP-60089AZ1
  • MSPP-60089AZ
  • Antigen Name:
  • CD184 (CXCR4)
  • CD184 (CXCR4)
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • APC (Allophycocyanin)
  • APC (Allophycocyanin)
  • Clone:
  • 12G5
  • 12G5
  • Reactivity:
  • Baboon, African green monkey, Cynomolgus monkey, Sooty mangabey, Rhesus monkey, Chimpanzee, Human
  • Baboon, African green monkey, Cynomolgus monkey, Sooty mangabey, Rhesus monkey, Chimpanzee, Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 7852
  • 7852
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa

Specifications

Frequently Bought Together