Hamster (Armenian) monoclonal IgG1 antibody against mouse CD3e, PerCP-Cy5.5-conjugated.
- T cells
The 145-2C11 antibody reacts with the ~20 kDa CD3ε subunit of the mouse T cell receptor (TCR)/CD3 complex, which is expressed on the surface of circulating mature T cells and NKT cells, and variably on thymocytes. A majority of T cell neoplasms also express CD3. The CD3 complex, which is assembled from combinations of CD3γ, δ, ε, η, and ζ subunits, associates non-covalently with the TCR and is involved in transducing antigen recognition signals into the cytoplasm of T cells and in regulating the cell surface expression of the TCR. Activation of T cells by the TCR involves the cytoplasmic tails of the CD3 subunits, which are structurally related type 1 transmembrane proteins and members of the immunoglobulin superfamily. Mutations in the CD3 subunits have been associated with various immunodeficiency disorders including severed combined immunodeficiency (SCID). The 145-2C11 antibody has been used for in vitro functional (blocking and activation) assays and has been reported to block binding by the clone 17A2 antibody. The 145-2C11 antibody is not recommended for use with formalin-fixed, paraffin-embedded sections. This antibody clone has been verified for purity assessments of cells isolated with EasySep™ kits, including EasySep™ mouse T cell isolation kit (Catalog #19851) and EasySep™ mouse CD90.2 positive selection kit II (Catalog #18951).
Type: Anti-CD38 Mouse Monoclonal Antibody [clone: HIT2] (PE)
Antigen: CD38
Clonality: Monoclonal
Clone: HIT2
Conjugation: PE
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Chimpanzee, Horse, Human
- Catalog No:
- MSPP-60014PE
- MSPP-60014PE1
- Antigen Name:
- CD38
- CD38
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- PE (Phycoerythrin)
- PE (Phycoerythrin)
- Clone:
- HIT2
- HIT2
- Reactivity:
- Horse, Chimpanzee, Human
- Horse, Chimpanzee, Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 952
- 952
- Isotype:
- IgG1 kappa
- IgG1 kappa