Order Entry
United States
ContactUsLinkComponent
Anti-CD45.2 Mouse Monoclonal Antibody [clone: 104] (APC)
Anti-CD45.2 Mouse Monoclonal Antibody [clone: 104] (APC)
Catalog # MSPP-100-1598
undefined
Anti-CD45.2 Mouse Monoclonal Antibody [clone: 104] (APC)
Catalog # MSPP-100-1598
Supplier Number:  100-1598
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Antigen Name:
    CD45.2
  • Clonality:
    Monoclonal
  • Clone:
    104
  • Conjugation:
    APC (Allophycocyanin)
  • Host:
    Mouse
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Mouse
  • Size:
    100 µg
  • Environmentally Preferable:
  • Gene ID:
    19264
  • Cat. No.:
    MSPP-100-1598

Specifications

About this item

Mouse monoclonal antibody against mouse CD45.2, APC-conjugated.

  • Granulocytes
  • Subsets
  • Hematopoietic stem
  • Progenitor cells
  • Lymphocytes
  • Myeloid cells

This monoclonal antibody (clone 104) reacts with mouse alloantigen cluster of differentiation 45.2 (CD45.2), also known as Ly5.2 and leukocyte common antigen, a 180 -240 kD type I transmembrane glycoprotein. CD45.2 is an allelic form of CD45 expressed by all hematopoietic cells except mature erythrocytes and platelets in mouse strains 129, A, AKR, BALB/c, C3H/He, C57BL/6, C58, CBA, DBA/1, and DBA/2. Use this recombinant monoclonal antibody in applications requiring detection of CD45.2, such as flow cytometry (FC). This antibody clone has been verified for purity assessment of cells isolated with EasySep™ mouse CD45 positive selection kit (Catalog #18945) or EasySep™ mouse TIL (CD45) positive selection kit (Catalog #100-0350). Clone 104 does not react with leukocytes that express the CD45.1 alloantigen. CD45 is a member of the protein tyrosine phosphatase (PTP) family and contains two catalytic domains in the intracellular (COOH) domain, which regulate cellular signaling. The extracellular domains are highly variable due to alternative splicing among exons 4, 5, and 6 as well as differential glycosylation. These CD45 isoforms correlate to specific cell types as well as specific activation and maturation states of immune cells. This monoclonal antibody has been used extensively in adoptive cell transfer and bone marrow transplantation studies in mice, allowing for the differentiation of CD45.2+ and CD45.1+ cells.

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Type: Anti-CD45.2 Mouse Monoclonal Antibody [clone: 104] (APC)
Antigen: CD45.2
Clonality: Monoclonal
Clone: 104
Conjugation: APC
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Mouse