About this item
T4 recombination mediator protein, UvsY plays important roles in homologous recombination (HR).
- During HR, the UvsX recombinase competes with the prebound single-stranded binding protein gp32 for DNA-binding sites
- UvsY stimulates this filament nucleation event
UvsY stimulates the DNA-dependent ATPase activity of UvsX, lowers the critical concentration of UvsX that is required for activity, and promotes strand exchange.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Conjugation:His
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Bacteriophage lambda
- Protein Synonyms:UvsY
- Protein/Peptide Name:T4 UvsY protein