Saturix C30 Polyester/Cellulose Wipes
Supplier: FG CLEAN WIPES TE
Certificates
About this item
These polyester/cellulose wipes offer the essential benefits of a non-woven wipe, high absorbency with aqueous liquids, dependable durability, and value. The exacting manufacturing process combines versatility, economy, and cleanliness.
- Hydroentangled blend of 55% cellulose and 45% polyester fabric (64 g/m²)
- Super absorbent
- Extremely low particle generation
- No chemical binders
Economical, general-purpose wipes offer extremely low particle generation with no chemical binders. Manufactured with a standard weight, hydroentangled blend of 55% cellulose and 45% polyester, C30 wipes are highly absorbent and residue-free, with low NVRs and ion extractalbes. Recommended for use in ISO 5 to 8 controlled environments.
Frequently Bought Together