Order Entry
United States
Orders LinkContactUsLinkComponent
Intracellular Permeabilization Buffer (10X)

Intracellular Permeabilization Buffer (10X)

Reagent for the permeabilization of fixed cells, 150 ml
Catalog # MSPP-100-1451
Marketsource
Supplier #:  100-1451
Intracellular Permeabilization Buffer (10X)
Catalog # MSPP-100-1451
Marketsource
CAS Number:  
Intracellular Permeabilization Buffer (10X)
Intracellular Permeabilization Buffer (10X)

Reagent for the permeabilization of fixed cells, 150 ml

CAS Number:  
Price
$190.00
Log in for availability
Each(1/Itm/Bx)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Ideal for the permeabilization of fixed cells during labeling of cytokines and other cytoplasmic antigens.

Intracellular Permeabilization Buffer (10X) is uniquely formulated for optimal resolution and to reduce background and is suitable for use with fluorochrome-conjugated antibodies for flow cytometry.

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifications

Specifications

  • Description:
    Reagent for the permeabilization of fixed cells
  • Size:
    150 ml
  • Application:
    Immunology

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.