Order Entry
United States
Orders LinkContactUsLinkComponent
Fixation Buffer

Fixation Buffer

Ready-to-use buffer to preserve cells prior to intracellular staining or flow cytometry, 100 ml
Catalog # MSPP-100-1450
Marketsource
Supplier #:  100-1450
Fixation Buffer
Catalog # MSPP-100-1450
Marketsource
CAS Number:  
Fixation Buffer
Fixation Buffer

Ready-to-use buffer to preserve cells prior to intracellular staining or flow cytometry, 100 ml

CAS Number:  
Price
$140.31
Log in for availability
Each(1/Itm/Bx)
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Ideal for fixing cells prior to permeabilization, Fixation Buffer can be used with intracellular permeabilization buffer (10X) to conduct intracellular staining for flow cytometry.

This fixation buffer can also be used to preserve fluorescently labeled cells prior to analysis by flow cytometry.

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifications

Specifications

  • Description:
    Ready-to-use buffer to preserve cells prior to intracellular staining or flow cytometry
  • Size:
    100 ml
  • Application:
    Immunology

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.