Orders LinkContactUsLinkComponent
Ammonium Chloride Solution
Ammonium Chloride Solution
Marketsource
Ammonium Chloride Solution
Marketsource
Certificates
Ammonium Chloride Solution
Ammonium Chloride Solution
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Ammonium chloride solution is recommended for the lysis of red blood cells (RBCs) in preparations of human and mouse peripheral blood, spleen or bone marrow cells.
It is buffered and optimized for gentle lysis of erythrocytes, with minimal effect on leukocytes. It does not contain a fixative agent, therefore leukocytes are viable following RBC lysis.
Species: Human, mouse, non human primate, rat and others.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Product Family Options
Product Information
  • Volume
    Availability
    Price
  • MSPP-07800
    Reagent for lysis of red blood cells
    Reagent for lysis of red blood cells
    100 ml
    Log in for availability
    Log in for availability
    Each
    Each - $54.08
    $54.08
    Each
       $54.08
    Supplier Number: 07800
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    Marketsource
    Marketsource
  • MSPP-07850
    Reagent for lysis of red blood cells
    Reagent for lysis of red blood cells
    500 ml
    Log in for availability
    Log in for availability
    Each
    Each - $190.00
    $190.00
    Each
       $190.00
    Supplier Number: 07850
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    Marketsource
    Marketsource