ALDEFLUOR™ Assay Buffer
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Use ALDEFLUOR™ Assay Buffer to optimize the detection of aldehyde dehydrogenase-bright (ALDHbr) cells and maximize fluorescent signal stability during the ALDEFLUOR™ assay.
ALDEFLUOR™ Assay Buffer is recommended for use in all procedures performed after ALDH staining to inhibit the active efflux of the ALDEFLUOR™ reaction product and to avoid loss in the assay signal.
Application: Cancer, epithelial cell biology, neuroscience, stem cell biology
Species: Human, mouse, non human primate, rat and others.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Specifications
Specifications
- Description:Additional assay buffer for use with the ALDEFLUOR™ kit
- Size:55 ml
- Cat. No.:MSPP-01702
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



