Order Entry
United States
ContactUsLinkComponent
Hank's Balanced Salt Solution (HBSS), Modified (Without Ca⁺⁺ and Mg⁺⁺)
Hank's Balanced Salt Solution (HBSS), Modified (Without Ca⁺⁺ and Mg⁺⁺)
Catalog # MSPP-37250
CAS Number:  
undefined
Hank's Balanced Salt Solution (HBSS), Modified (Without Ca⁺⁺ and Mg⁺⁺)
Catalog # MSPP-37250
Supplier Number:  37250
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Hank's Balanced Salt Solution (HBSS) without calcium and magnesium
  • Cat. No.:
    MSPP-37250
  • Volume:
    500 ml

Specifications

About this item

Hank's Balanced Salt Solution (HBSS) can be used for diluting, washing, irrigating or as a transporting solution for cell or tissue culture. It provides a buffering system to maintain the physiological pH range and osmotic balance of culture media, and provides cells with a source of water and essential inorganic ions and a carbohydrate as an energy source.

Selection of suitable nutrient medium is dependent on type of cell, culture conditions, and degree of chemical definition required for the cell culture application.

Species: Human, mouse, non human primate, rat and others.

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').