Orders LinkContactUsLinkComponent
Corning® Lambda™ Plus 12-Channel Pipettor
Corning® Lambda™ Plus 12-Channel Pipettor
Lambda™ Plus 12-channel pipettor, 20 - 200 μl
Catalog # MSPP-38064
Marketsource
Supplier #:  38064
Corning® Lambda™ Plus 12-Channel Pipettor
Catalog # MSPP-38064
Marketsource
Corning® Lambda™ Plus 12-Channel Pipettor
Corning® Lambda™ Plus 12-Channel Pipettor
Lambda™ Plus 12-channel pipettor, 20 - 200 μl
Price
$2,018.38
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
Measure and transfer liquids precisely and safely using Corning® Lambda™ Plus multi-channel pipettors.
  • Autoclavable, UV-resistant 12-channel micropipette
  • Universal fit for all common pipette tip brands
  • Low pipetting force
  • Adjustable tip ejector
Fully autoclavable and UV-resistant, this 12-channel, 200 μl volumetric instrument is designed for a universal fit with all common brands of pipette tips, including Corning® filtered pipette tips. Each multi-channel pipettor has a smooth plunger movement, low pipetting force, and an adjustable tip ejector. Volume can be adjusted using the thumb wheel or plunger button and a locking mechanism can be used to prevent accidental volume changes. It is supplied with a key for easy in-house calibration and has been designed to maintain calibration after repeated use and autoclaving.
 : Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Specifications
  • Volume:
    20 - 200 μl
  • Description:
    Lambda™ Plus 12-channel pipettor