ContactUsLinkComponent
EasyPlate™ EasySep™ Magnet
EasyPlate™ EasySep™ Magnet
Catalog # MSPP-18102
undefined
EasyPlate™ EasySep™ Magnet
Catalog # MSPP-18102
Supplier Number:  18102
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    EasyPlate™ EasySep™ magnet
  • Environmentally Preferable:
  • Cat. No.:
    MSPP-18102
Specifications
About this item
The EasyPlate™ EasySep™ Magnet is designed for cell separation procedures using selected EasySep™ reagents when processing a small number of cells (up to 2×10⁷ cells per well).
  • Immunomagnetic column-free magnet
  • Designed to hold a round-bottom, non-tissue culture treated 96-well plate
The EasyPlate™ EasySep™ magnet generates a high-gradient magnetic field that is strong enough to separate cells labeled with EasySep™ D magnetic particles, without the use of columns. The EasyPlate™ EasySep™ magnet is designed to hold a round-bottom, non-tissue culture treated 96-well plate.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').