SteriWare Swabbing Templates
New Product
Swabbing templates ensure a consistent swabbing area for cleaning validation purposes.
- Choice of sterile or non sterile
- Ensure consistent validation results
- Individually bagged to reduce contamination
These swabbing templates are made in a cleanroom and come individually bagged to help reduce the risk of contamination.
Frequently Bought Together