Orders LinkContactUsLinkComponent
Dipping Bottles
Dipping Bottles
Dipping Bottles
Certificates
Dipping Bottles
6 Options Available Below
Product Details & Documents
Dipping bottles are excellent for sampling in open water, as well as in tanks and drums.
  • Wide range available
  • Stainless steel 304 or brass construction
  • Customisable
Sampling liquids at depth is easy with these dipping bottles. Available in either 304 stainless steel or brass construction, they can take liquid samples from a variety of locations including open water kegs, drums, and wells.
Product Family Options
Product Information
  • Material
    Capacity
    Length
    Ø
    Availability
    Price
  • Brass
    300 ml
    162 mm
    75 mm
    Log in for availability
    Log in for availability
    Each
    Each - $266.00
    $266.00
    Each
       $266.00
    Supplier Number: 5332B-300
  • Brass
    500 ml
    240 mm
    75 mm
    Log in for availability
    Log in for availability
    Each
    Each - $292.00
    $292.00
    Each
       $292.00
    Supplier Number: 5332B-500
  • Brass
    1000 ml
    300 mm
    75 mm
    Log in for availability
    Log in for availability
    Each
    Each - $318.00
    $318.00
    Each
       $318.00
    Supplier Number: 5332B-1000
  • Stainless steel 304
    300 ml
    153 mm
    62 mm
    Log in for availability
    Log in for availability
    Each
    Each - $436.00
    $436.00
    Each
       $436.00
    Supplier Number: 5332S-300
  • Stainless steel 304
    500 ml
    207 mm
    62 mm
    Log in for availability
    Log in for availability
    Each
    Each - $483.00
    $483.00
    Each
       $483.00
    Supplier Number: 5332S-500
  • Stainless steel 304
    1000 ml
    325 mm
    77 mm
    Log in for availability
    Log in for availability
    Each
    Each - $529.00
    $529.00
    Each
       $529.00
    Supplier Number: 5332S-1000