Measuring Scoops
Supplier: SAMPLING SYSTEMS LTD
Jump to:
About this item
These measuring scoops made from high quality 316 stainless steel are ideal for scooping out fixed volumes of powder.
- Crevice-free
- Polished
5 Options Available Below
Product Details & Documents
These measuring scoops made from high quality 316 stainless steel are ideal for scooping out fixed volumes of powder.
- Crevice-free
- Polished
Product Family Options
Product Information
- CapacityLengthØAvailabilityPrice
- Supplier Number: 1900A-01
- Supplier Number: 1900A-05
- Supplier Number: 1900A-10
- Supplier Number: 1900A-25
- Supplier Number: 1900A-50