Order Entry
United States
ContactUsLinkComponent
MesenCult™ ACF Freezing Medium
MesenCult™ ACF Freezing Medium
Catalog # MSPP-05490
CAS Number:  
undefined
MesenCult™ ACF Freezing Medium
Catalog # MSPP-05490
Supplier Number:  05490
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    MesenCult™-ACF Freezing Medium, 50 ml
  • Environmentally Preferable:
  • Cat. No.:
    MSPP-05490
  • Format:
    Human; animal component-free, serum-free

Specifications

About this item

Animal component-free MSC cryopreservation medium.

  • Serum-free and animal component-free
  • Reproducibly high recovery rates
  • Optimized for MSCs previously cultured in MesenCult™-ACF Plus medium or MesenCult™ medium
  • Preserves human MSC multipotency and expansion capacities
  • Convenient, ready-to-use format

MesenCult™-ACF Freezing medium is a defined, serum-free, and animal component-free medium for the cryopreservation of mesenchymal stem cells (MSCs). This complete and ready-to-use medium is recommended for human MSCs previously cultured in MesenCult™-ACF Plus medium or MesenCult™ medium (MesenCult™ proliferation kit). Frozen human MSCs should be stored at –135 °C (liquid nitrogen) or colder.

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').