About this item
Leukemia inhibitory factor (LIF) is an interleukin 6 class cytokine that regulates a broad variety of developmental functions. After LIF binds to the LIF receptor (LIFR), LIFR associates with gp130 and activates JAK/STAT and MAPK signaling (Auernhammer and Melmed; Suman et al.). LIFR activation of STAT3 is essential for maintaining the mouse embryonic stem cell phenotype (Niwa et al.). Produced by the endometrium, LIF plays an important autocrine and paracrine role in implantation by regulating proliferation, invasion, and differentiation of trophoblasts following blastocyst attachment (Auernhammer and Melmed; Suman et al.). Human LIF can be used for the maintenance of mouse embryonic stem cells; however, mouse LIF cannot bind to the human receptor, thus rendering mouse LIF inactive (Dahéron et al.). LIF is produced by CD4+ and activated regulatory T cells, and promotes Foxp3 expression, while repressing Th17 lineage-specific genes (Metcalfe). LIF is also secreted by mesenchymal stromal cells, where it supports hematopoiesis and immune modulation (Nasef et al.).
Specifications
- Protein/Peptide Type:Recombinant
- Species:Mouse
- Protein/Peptide Name:LIF
- Purity:≥95%