About this item
Fibroblast growth factor 8B (FGF-8B) is a member of the fibroblast growth factor (FGF) family and is an isoform of FGF-8. Cytokines in the FGF family possess broad mitogenic and cell survival activities (Folkman andamp; Klagsbrun; Kimelman andamp; Kirschner) and are involved in a variety of biological processes, including cell proliferation, differentiation, survival, and apoptosis (Folkman andamp; Klagsbrun; Klagsbrun; Rifkin andamp; Moscatelli). FGF-8B signals through FGF receptors (FGFRs) to activate PI3K and MAPK pathways. FGF-8B is broadly associated with mitogenic and cell survival activities, and regulates gastrulation, epithelial-mesenchymal transition, and later on mesenchymal to epithelial differentiation during embryonic development. FGF-8B has also been found in peripheral blood leukocytes and healthy bone marrow samples (Mattila andamp; Härkönen). FGF-8B has mitogenic effects on somatic cells in the germinal epithelium and is expressed in adult mouse ovarian cells and tissues, which suggests that it regulates maturation of oocytes and seminiferous epithelium in testis (Valve et al.).
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Protein/Peptide Type:Recombinant
- Species:Human
- Protein/Peptide Name:FGF-8B
- Purity:>95%