RANTES (regulated upon activation, normal T cell expressed and secreted), also known as CCL5, is a member of the CC family of chemokines and is able to recruit leukocytes to sites of inflammation (Schall et al.). RANTES is secreted by T lymphocytes, macrophages, platelets, synovial fibroblasts, tubular epithelium, and certain types of tumor cells (Aldinucci and Colombatti; Soria and Ben-Baruch). This chemokine exerts its effect by interacting with the chemokine receptors CCR1, CCR3, CCR4, and CCR5. RANTES plays an active role in recruiting a variety of leukocytes into inflammatory sites, including T cells, macrophages, eosinophils, and basophils. In collaboration with certain cytokines that are released by T cells such as IL-2 and IFN-γ, RANTES also induces the activation and proliferation of NK cells to generate CC chemokine-activated killer cells, which are highly cytolytic (Lv et al.; Maghazachi et al.). It has been shown that RANTES produced by CD8+ T cells inhibits HIV infection of primary human peripheral blood mononuclear cells (Appay and Rowland-Jones; Cocchi et al.).
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Protein/Peptide Type:Recombinant
- Species:MouseRat
- Protein/Peptide Name:RANTES (CCL5)
- Purity:≥95%