About this item
Macrophage inflammatory protein-1 beta (MIP-1 beta), also known as CCL4, is a member of the CC family of chemokines and is most closely related to CCL3 (MIP-1 alpha). Cellular sources of MIP-1 beta include activated leukocytes (monocytes and T and B cells), brain endothelial cells, and smooth muscle cells (Lukacs et al.; Menten et al.). MIP-1 beta, MIP-1 alpha, and RANTES have been shown to be major HIV-suppressive factors, possibly through the interactions of these chemokines with the receptor CCR5 on CD4+ T cells, which is also a major receptor for HIV entry into CD4+ T cells (Cocchi et al.; Menten et al.). MIP-1 beta attracts a variety of immune cells to sites of microbial infection. In addition to its chemotactic functions, MIP-1 beta induces the release of proinflammatory cytokines, mast cell degranulation, and NK cell activation (Schall et al.). In mice, recruitment of regulatory T cells to B cells and antigen-presenting cells by MIP-1 beta plays a central role in the initiation of T cell and humoral responses, and the depletion of regulatory T cells or MIP-1 beta results in deregulated humoral responses and production of autoantibodies (Bystry et al.).
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Protein/Peptide Type:Recombinant
- Species:Mouse
- Protein/Peptide Name:MIP-1 beta (CCL4)
- Purity:≥95%