Roto-Rock Genie Rotator Rocker, 230 V
Catalog # 77574-962
Supplier: Scientific Industries
New Product
Irregular Voltage
Specifications
- Rotation Speed (min-1):1 - 70 cycles/min; 1 - 35 rpm
- Temperature:0...38 °C
- Description:Roto-Rock Genie, without plug, 230 V (export)
- Electrical:Input: 90 - 264 V AC, 50/60 Hz
- Shipping Weight:10.2 kg (22.5 lbs.)
- Max. Load:4.5 kg (10 lbs.)
- Platform Size:320×250 mm
- Dimensions:400×300×380 mm
- Timer:1 s - 99 h, 59 min, 59 s, or continous
- Weight:9.7 kg (21.3 lbs.)
- Rocking Angle:±1 - 25° from horizontal
- Cat. No.:77574-962
Specifications
About this item
Programmable combination rotator rocker with digital touch screen, featuring magnetic platform and clip plate system.
- 230 V model
- Intuitive touch screen control makes changing program settings easy
- Setting the tilt angle and speed control provides thorough mixing while eliminating spills when using open containers in rocking mode
- The two-sided magnetic platform holds virtually any type of tube and other sample vessels used in the laboratory when using the GENIE clip plates (sold separately)
- Compact design saves valuable bench space in the laboratory
- Designed for use in cold rooms, incubators (non-CO₂), or at ambient temperature
- 2 year warranty
Programable all-in-one benchtop GENIE Rocker/Rotator or cycle from Rock to Rotate. Digital set/control for rpm, total time, cycle time. Features 'pause/resume' button allowing the operator to add reagents to a sample during processing.
Rocking tray size: 300×300 mm
Power: DC 24 V, 2.5 A
Certifications: CE certified.
Delivery: Supplied with magnetic platform and non-slip rocking tray. Accessories must be ordered separately: GENIE clip plate systems designed to handle a variety of vessels from PCR tube, vials, bottles and sealed bags.
Caution: 230 V model. Corresponding model with voltage suitable for use in US (77574-960) is also available, please see separate product page.