Persephin is a neurotrophic factor that belongs to the glial cell line-derived neurotrophic factor (GDNF) family. Persephin shares a large degree of structural similarity to GDNF, artemin, and neurturin, and has overall neuroprotective activity. Persephin signals through GRFα4 (glycosylphosphatidylinositol (GPI)-linked GDNF receptor family member) which signals through the receptor tyrosine kinase RET. Unlike GDNF and neurturin, persephin only promotes the growth and survival of central dopaminergic and motor neurons, but not peripheral neurons (Milbrandt et al.). In vitro persephin only promotes survival of neurons that co-express GPI-linked GRFα4 and RET (Enokido et al.; Lindahl et al.). Mice lacking persephin showed increased cell death after cerebral ischemia, however administration of persephin before ischemia dramatically reduced neuronal cell death (Tomac et al.). This product is animal component-free.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Protein/Peptide Type:Recombinant
- Species:Human
- Protein/Peptide Name:Persephin, ACF
- Purity:≥95%