Fibroblast growth factor 7 (FGF-7) is a member of the FGF family, and acts exclusively through a subset of FGF receptor isoforms expressed predominantly by epithelial cells (Finch and Rubin). FGF-7 seems to act specifically on epithelial cells and stimulates proliferation, migration, and differentiation of these cells, and also participates in epithelial protection and repair both in vitro and in vivo (Finch and Rubin; Werner). In contrast, FGF-7 is produced solely by cells of mesenchymal origin, and functions as a paracrine mediator of mesenchymal-epithelial communication (Rubin et al.). FGF-7 has also been shown to supplement several wound-healing properties of bioengineered skin (Erdag et al.) and to induce autophagy in human keratinocytes (Belleudi et al.). Additionally, FGF-7 has a role in the differentiation of pluripotent stem cell to endodermal pancreatic-like insulin-producing cells and thymic epithelial cells (Inami et al.; Niu et al.). This product is animal component-free.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Protein/Peptide Type:Recombinant
- Species:Human
- Protein/Peptide Name:FGF-7, ACF
- Purity:≥90%