About this item
Fractalkine (CX3CL1) is a unique chemokine belonging to the CX3C family, and is characterized by a C-X3-C cysteine motif within the chemokine domain, near the amino terminus of the protein (Bazan et al.). The chemokine domain is connected to an extended mucin-like stalk, followed by a transmembrane region, and a C-terminal intracellular domain (Imai et al.; Jones et al.). The protein signals through interaction with a single receptor, CX3CR1, expressed on monocytes, natural killer cells, T cells, microglia, and smooth muscle cells. Fractalkine is upregulated in endothelial cells by inflammatory signals and is synthesized as a membrane-bound molecule that mediates cell migration and adhesion (White and Greaves). Cleavage at the base of the stalk by metalloproteinases generates a soluble chemokine, which functions as a potent chemoattractant of target cells (Garton et al.; Apostolakis and Spandidos). Fractalkine has been implicated in pathology of inflammatory diseases, such as atherosclerosis and other vascular diseases, and has anti-apoptotic functions (White and Greaves).
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Protein/Peptide Type:Recombinant
- Species:Human
- Protein/Peptide Name:Fractalkine (CX3CL1)
- Purity:≥95%