Order Entry
United States
ContactUsLinkComponent
Human Recombinant BAFF
Human Recombinant BAFF
Certificates
Human Recombinant BAFF

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Species:
    Human
  • Protein/Peptide Name:
    BAFF
  • Purity:
    95%

Specifications

Frequently Bought Together