Order Entry
United States
ContactUsLinkComponent
Human Recombinant CD40 Ligand
Human Recombinant CD40 Ligand
Certificates
undefined
Human Recombinant CD40 Ligand

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Species:
    Human
  • Protein/Peptide Name:
    CD40 Ligand
  • Purity:
    ≥95%

Specifications

Frequently Bought Together