About this item
Stromal cell-derived factor 1 alpha (SDF-1α) is a member of the CXC group of chemokines that binds to the G-protein coupled receptor, CXCR4, to regulate migration, proliferation, differentiation, and survival of many cell types including hematopoietic stem cells, B cells, and T cells. It is produced by bone marrow stromal cells, osteoblasts, endothelial cells, and neuronal cells. SDF-1α was first identified as the pre-B-cell growth-stimulating factor (PBSF) in the mouse bone marrow-derived stromal cell line, PA6, in the growth of B cell precursors (Hayashi et al.). SDF-1α primarily regulates cell motility during development and adulthood, including the homing of hematopoietic stem cells and neutrophils to fetal bone marrow during ontogeny (Ara et al. 2003a) and the recruitment of endothelial progenitor cells from bone marrow during angiogenesis in adulthood (Zheng et al.). In addition to its role in hematopoiesis, the SDF-1α/CXCR4 signaling pathway is also essential for the homing of primordial germ cells to gonads (Ara et al. 2003b), the migration of granule cells in the cerebellum during neurogenesis (Zou et al.), and the migration of breast cancer cells to sites of metastasis (Muller et al.).
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Protein/Peptide Type:Recombinant
- Species:Mouse
- Protein/Peptide Name:SDF-1 alpha (CXCL12)
- Purity:≥95%