Order Entry
United States
Orders LinkContactUsLinkComponent
Human Recombinant NGF-beta

Human Recombinant NGF-beta

Marketsource
Human Recombinant NGF-beta
Marketsource
Certificates
Human Recombinant NGF-beta

2 Options Available Below

  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Nerve growth factor (NGF)-beta is a prototypical member of the neurotrophin family and has a role in the survival and growth of neural cells, regulating cell growth, promoting differentiation into neurons, and neuron migration. The beta subtype of NGF is biologically active in comparison to the alpha-2 and gamma-2 subtypes. NGF-beta in its secreted form can bind to tyrosine kinase A (TrkA) receptor with high affinity and to p75 (NTR) with low affinity (Levi and Alemà; Sofroniew et al.). NGF has been shown to possess pro-inflammatory and pro-fibrogenic properties (Micera et al.). It has also been shown that overexpression of NGF-beta promotes differentiation of bone marrow mesenchymal stem cells into neurons through regulation of AKT and MAPK pathways (Yuan et al.).

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • MSPP-78092
  • MSPP-780921006
  • Protein/Peptide Name:
  • NGF-beta
  • NGF-beta
  • Protein/Peptide Type:
  • Recombinant
  • Recombinant
  • Species:
  • Human
  • Human
  • Purity:
  • ≥95%
  • ≥95%

Specifications

Product Family Options

Product Information

  • Size
    Availability
    Price
  • 20 µg
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $135.92
    $135.92
    Each(1/Itm/Bx)
       $135.92
    Minimum order quantity is 1

    Supplier Number: 78092
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    MSPP-78092
    Protein/Peptide Name
    NGF-beta
    Protein/Peptide Type
    Recombinant
    Species
    Human
    Purity
    ≥95%
    Marketsource
  • 100 µg
    Log in for availability
    Log in for availability
    Each(1/Itm/Bx)
    Each(1/Itm/Bx) - $331.77
    $331.77
    Each(1/Itm/Bx)
       $331.77
    Minimum order quantity is 1

    Supplier Number: 78092.1
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    Marketsource
    More
    Specifications
    Cat. No.
    MSPP-780921006
    Protein/Peptide Name
    NGF-beta
    Protein/Peptide Type
    Recombinant
    Species
    Human
    Purity
    ≥95%
    Marketsource
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.