Order Entry
United States
ContactUsLinkComponent
Mouse Recombinant EGF
Mouse Recombinant EGF
Certificates
undefined
Mouse Recombinant EGF

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Species:
    Mouse
  • Protein/Peptide Name:
    EGF
  • Purity:
    ≥95 %

Specifications

Frequently Bought Together