To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
- Protein/Peptide Type:Recombinant
- Species:Human
- Size:25 µg
- Protein/Peptide Name:IL-8 (CXCL8)
- Purity:≥95 %
- Cat. No.:MSPP-780841006
Interleukin-8 (IL-8) is a member of the CXC subfamily of chemokines and is produced by leukocytic cells (monocytes, T cells, neutrophils, and natural killer cells) and non-leukocytic somatic cells (endothelial cells, fibroblasts, and epithelial cells), with the most prominent source being monocytes and macrophages. Its production is induced by inflammatory stimuli, such as IL-1. IL-8, also known as CXCL8, activates neutrophils inducing chemotaxis, exocytosis, and the respiratory burst (Baggiolini and Clark-Lewis; Mukaida). IL-8 is considered one of the most potent neutrophil chemoattractants in inflammation and binds to two different chemokine receptors on leukocytes: the G protein-coupled receptors CXCR1 and CXCR2 (Hoffmann et al.; de Oliveira et al.). IL-8 has angiogenic effects on human intestinal microvascular endothelial cells in vitro that are mediated by CXCR2 (Heidemann et al.). IL-8 is reported to promote breast cancer progression by increasing cell invasion, angiogenesis, and metastasis and has been reported to be involved in regulating breast cancer stem-like cells (Singh et al.). IL-8 also has proangiogenic properties in inflammatory diseases of the conjunctiva, cornea, iris, retina, and orbit (Ghasemi et al.). It was also shown that a major T cell effector function in human newborns is IL-8 production, which has the potential to activate antimicrobial neutrophils and gamma/delta T cells (Gibbons et al.). A variety of human pathogens, such as HIV and Mycobacterium tuberculosis, have been shown to induce IL-8 production by monocytes and macrophages (Friedland et al.; Meddows-Taylor et al.).