Specifications
- Size:1 unit
- Protein/Peptide Name:EBV (HLA Class I Control) Peptide Pool
- Purity:≥95%
- Cat. No.:MSPP-100-1391
Specifications
About this item
Stimulating T cells with EBV (HLA Class I Control) peptide pool releases downstream cytokines and upregulates activation markers, enabling antigen-specific T cells to be detected or isolated for analysis. EBV (HLA Class I Control) Peptide Pool is a lyophilized mixture of 26 peptides from Epstein-Barr virus (EBV), and consists of defined HLA class I-restricted T cell epitopes. Viral peptide pools are useful for a broad range of applications, including vaccine development, immunological research, and diagnostic assay development. For consistency and reproducibility across your applications, CMV peptide pool from STEMCELL has a purity ≥95%.