Order Entry
United States
ContactUsLinkComponent
Human Recombinant Complement Factor D, His Tag
Human Recombinant Complement Factor D, His Tag
Catalog # MSPP-100-1330
CAS Number:  
undefined
Human Recombinant Complement Factor D, His Tag
Catalog # MSPP-100-1330
Supplier Number:  100-1330
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Species:
    Human
  • Size:
    50 µg
  • Protein Synonyms:
    #N/A
  • Protein/Peptide Name:
    Complement Factor D
  • Purity:
    ≥94%
  • Cat. No.:
    MSPP-100-1330

Specifications

About this item

Complement factor D is a component of the alternative pathway of the complement system and part of the innate immune system, playing a vital role in the initiation and amplification of complement activation, in order to defend against infection (Barratt and Weitz). A serine protease belonging to the S1 peptidase family, complement factor D is secreted by adipocytes into circulating blood, and is also expressed by macrophages and monocytes (White et al.). In the initiation phase of the complement pathway, complement factor D cleaves complement factor B (bound to component C3) to produce a complex known as C3 convertase. During the amplification phase, complement factor D cleaves complement factor B (bound to component C3b) to produce the C3bBb convertase, and is involved in the propagation of complement activation. In addition to its immunological role, complement factor D is involved in other physiological processes, such as the efficient clearing of damaged cell debris by phagocytes following acute liver injury (Cresci et al.). Complement factor D deficiency is associated with an increased susceptibility to pathogens like Neisseria meningitidis (Biesma et al.). This protein contains a His-residue tag at the carboxyl end of the polypeptide chain. For consistency and reproducibility across your applications, complement factor D from STEMCELL comes lyophilized with ≥94% purity, and is verified by LAL analysis to ensure endotoxin levels are ≤1.0 EU/μg protein.

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').