Order Entry
United States
ContactUsLinkComponent
SARS-CoV-2 (VME1) Peptide Pool
SARS-CoV-2 (VME1) Peptide Pool
Catalog # MSPP-100-0648
CAS Number:  
undefined
SARS-CoV-2 (VME1) Peptide Pool
Catalog # MSPP-100-0648
Supplier Number:  100-0648
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Size:
    1 unit
  • Protein/Peptide Name:
    SARS-CoV-2 (VME1) Peptide Pool
  • Purity:
    Average 70%
  • Cat. No.:
    MSPP-100-0648

Specifications

About this item

The SARS-CoV-2 (VME1) peptide pool is a lyophilized mixture of 53 peptides from the SARS-CoV-2 membrane protein VME1. VME1 is a structural protein that is common to all coronaviruses, and it plays a critical role in viral assembly. The pool consists of 15-mer peptides with 11-amino-acid overlaps that cover amino acids 1 to 222 on VME1.

Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').