Order Entry
United States
ContactUsLinkComponent
Alpha 1-Antichymotrypsin ELISA
Alpha 1-Antichymotrypsin ELISA
Catalog # 101720-684
Supplier:  Alpco Diagnostics
CAS Number:  
Alpha 1-Antichymotrypsin ELISA
Catalog # 101720-684
Supplier:  Alpco Diagnostics
Supplier Number:  41-A1CHU-E01
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type:
    Sandwich
  • Conjugate ELISA:
    HRP
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Human
  • Target protein:
    Alpha 1-antichymotrypsin
  • Description:
    Alpha 1-antichymotrypsin ELISA
  • Size:
    1×96 wells
  • Sample Type:
    Plasma, serum
  • Detection Method:
    Colorimetric
  • Time to Results:
    1 h, 30 min
  • Detection Range:
    6.25 - 400 ng/ml
  • Sensitivity:
    6.25 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    101720-684
  • Supplier no.:
    41-A1CHU-E01

Specifications

About this item

The alpha 1-antichymotrypsin ELISA kit is a highly sensitive two-site enzyme linked immunoassay for measuring alpha 1-antichymotrypsin in biological fluids of humans.

In this assay the alpha 1-antichymotrypsin present in samples reacts with the anti-alpha 1-antichymotrypsin antibodies which have been adsorbed to the surface of polystyrene microtiter wells. After the removal of unbound proteins by washing, anti-CYT antibodies conjugated with horseradish peroxidase (HRP), are added. These enzyme-labeled antibodies form complexes with the previously bound CYT. Following another washing step, the enzyme bound to the immunosorbent is assayed by the addition of a chromogenic substrate, 3,3’,5,5’-tetramethylbenzidine (TMB). The quantity of bound enzyme varies directly with the concentration of CYT in the sample tested; thus, the absorbance, at 450 nm, is a measure of the concentration of CYT in the test sample. The quantity of CYT in the test sample can be interpolated from the standard curve constructed from the standards and corrected for sample dilution.

Caution: For research use only. Not for use in diagnostic procedures.