Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
- Assay duration:Multiple steps
- Assay Type:Competitive
- Conjugate ELISA:Peroxidase
- Format:Pre-coated
- Host:Goat
- Primary antibody reactivity:Human
- Target protein:Dopamine
- Description:Dopamine ELISA
- Size:1×96 wells
- Sample Type:Plasma, urine
- Detection Method:Colorimetric
- Time to Results:4 h
- Detection Range:10 - 2000 ng/ml
- Sensitivity:4.5 ng/ml
- Regulatory Status:RUO
- Cat. No.:101721-014
- Supplier no.:17-DOPHU-E01.1
The dopamine ELISA is an enzyme Immunoassay (ELISA) for the quantitative determination of dopamine in plasma and urine.
Dopamine is extracted by using a cis-diol-specific affinity gel, acylated and then derivatized enzymatically. The competitive ELISA kit uses the microtiter plate format. The antigen is bound to the solid phase of the microtiter plate. The derivatized standards, controls and samples and the solid phase bound analytes compete for a fixed number of antiserum binding sites. After the system is in equilibrium, free antigen and free antigen-antiserum complexes are removed by washing. The antibody bound to the solid phase is detected by an anti-rabbit IgG-peroxidase conjugate using TMB as a substrate. The reaction is monitored at 450 nm.