Order Entry
United States
ContactUsLinkComponent
Bi-Neph (Metanephrine and Normetanephrine) ELISA
Bi-Neph (Metanephrine and Normetanephrine) ELISA
Catalog # 101721-248
Supplier:  Alpco Diagnostics
CAS Number:  
Bi-Neph (Metanephrine and Normetanephrine) ELISA
Catalog # 101721-248
Supplier:  Alpco Diagnostics
Supplier Number:  17BINHUE02UFST
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type:
    Competitive
  • Conjugate ELISA:
    Peroxidase
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Human
  • Target protein:
    Metanephrine and normetanephrine
  • Description:
    Bi-Neph (metanephrine and normetanephrine) ELISA
  • Size:
    2×96 wells
  • Sample Type:
    Urine
  • Detection Method:
    Colorimetric
  • Time to Results:
    1 h, 45 min
  • Regulatory Status:
    RUO
  • Cat. No.:
    101721-248
  • Supplier no.:
    17BINHUE02UFST

Specifications

About this item

The Bi-Neph (Metanephrine and Normetanephrine) ELISA is for the quantitative determination of metanephrine and normetanephrine in urine.

First metanephrine (metadrenaline) and normetanephrine (normetadrenaline) are quantitatively acylated. The subsequent competitive ELISA kit uses the microtiter plate format. The antigen is bound to the solid phase of the microtiter plate. The acylated standards, controls, and samples and the solid phase bound analytes compete for a fixed number of antiserum binding sites. After the system is in equilibrium, free antigen and free antigen-antiserum complexes are removed by washing. The antibody bound to the solid phase is detected by an anti-rabbit IgG-peroxidase conjugate using TMB as a substrate. The reaction is monitored at 450 nm. Quantification of unknown samples is achieved by comparing their absorbance with a reference curve prepared with known standard concentrations. During the sample preparation, Metanephrine (Metadrenaline) is quantitatively acylated.

Caution: For research use only. Not for use in diagnostic procedures.