Order Entry
United States
ContactUsLinkComponent
Bi-CAT (Research) ELISA
Bi-CAT (Research) ELISA
Catalog # 101721-458
Supplier:  Alpco Diagnostics
CAS Number:  
Bi-CAT (Research) ELISA
Catalog # 101721-458
Supplier:  Alpco Diagnostics
Supplier Number:  17BCTHUE02RES
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type:
    Competitive
  • Conjugate ELISA:
    Peroxidase
  • Format:
    Pre-coated
  • Host:
    Goat
  • Primary antibody reactivity:
    Human
  • Target protein:
    Epinephrine and norepinephrine
  • Description:
    Bi-CAT (research) ELISA
  • Size:
    2×96 wells
  • Sample Type:
    Biological fluids, tissue homogenate
  • Detection Method:
    Colorimetric
  • Regulatory Status:
    RUO
  • Cat. No.:
    101721-458
  • Supplier no.:
    17BCTHUE02RES

Specifications

About this item

The Bi-CAT ELISA (Research) is a two-plate enzyme immunoassay for the quantitative determination of epinephrine and norepinephrine in biological fluids and tissue homogenate. Each catecholamine is measured independently on separate plates.

Epinephrine and norepinephrine (along with the third catecholamine, dopamine) act as hormones and neurotransmitters throughout the body. The release of both epinephrine and norepinephrine is associated with the 'fight or flight response' which in effect, prepares the body for action by increasing heart rate, blood pressure and the mobilization of fuels for use as energy. In turn, the actions of epinephrine and norepinephrine suppress non-emergency processes, such as digestion.

Caution: For research use only. Not for use in diagnostic procedures.