Order Entry
United States
ContactUsLinkComponent
Mouse Insulin ELISA Jumbo Pack (10-plates)
Mouse Insulin ELISA Jumbo Pack (10-plates)
Catalog # 101721-574
Supplier:  Alpco Diagnostics
CAS Number:  
Mouse Insulin ELISA Jumbo Pack (10-plates)
Catalog # 101721-574
Supplier:  Alpco Diagnostics
Supplier Number:  80-INSMS-E10
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    Insulin
  • Description:
    Mouse insulin ELISA jumbo pack (10-plate)
  • Size:
    10×96 wells
  • Sample Type:
    Plasma, serum
  • Detection Method:
    Colorimetric
  • Time to Results:
    2 h, 15 min
  • Detection Range:
    0.188 - 6.9 ng/ml
  • Sample Volume:
    10 µl
  • Sensitivity:
    0.06 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    101721-574
  • Supplier no.:
    80-INSMS-E10

Specifications

About this item

The mouse insulin ELISA Jumbo (10-plate) quantifies the concentration of insulin protein products in mouse I and mouse II proinsulin genes with just 10 µl of serum or plasma sample.

  • Low sample volume of 10 µl
  • Dynamic range of 0.188 to 6.9 ng/ml
  • Superior sensitivity of 0.06 ng/ml

Utilizing a dual-monoclonal antibody sandwich ELISA format, bi-level control set, and 96-well microplate comprising removable strips, the mouse insulin ELISA jumbo pack has the performance characteristics and flexibility necessary to confidently measure up to 40 samples in duplicate. The shelf-life of the components and the resealable microplate pouch allow for convenient storage for future use if an entire plate is not needed at one time.

Caution: For research use only. Not for use in diagnostic procedures.