Dynamic Weighing Kit
Catalog # 76183-636
Supplier: Mettler Toledo
Specifications
- Description:Dynamic weighing kit, used for S-platform
- Size:0.001 g/0.01 g platform
- Cat. No.:76183-636
Specifications
About this item
For Small, one and two place weighing pans on precision balances (XPR/XSR/XPE/XP/XS).
- Contaminant protection
- Easy to use
- For dynamic weighing with 4 L vessel
- Start to measure dynamically after quick and simple installation
- Not autoclavable