Charging stand for Eppendorf® Xplorer® Electronic Pipettors, Eppendorf
Supplier: Eppendorf
Certificates
About this item
This charging carousel holds up to three Eppendorf Xplorer® Series and one Repeater® Series Electronic Pipettes.
Use Charger Shell Adapter EPP-4880602000 to store and charge Eppendorf Repeater® M4, E3, E3x, stream and Xstream Electonic pipettors.
Delivery: AC Adapter & charging cable included.
Specifications
- Catalog No:
- 75874-588
- 75874-598
- Environmentally Preferable:
- Low Manufacturing Impact
- Environmental Certification:
- Renewable Energy Certificate, ISO 14001