Count-10™ Urinalysis Standardization Systems, Myers-Stevens Group
Supplier: Myers-Stevens Group
Certificates
About this item
This system provides a complete package for the standardization of microscopic urinalysis
It utilizes calibrated labware for urine collection and sample separation, precision test slides, and versatile quality control reagents for improved test accuracy and faster throughput. From specimen collection to microscopic sediment analysis, this system eliminates the procedure-dependent variables affecting a basic screening test.
Ordering information: The Count-10 Urine Collection System comes with 500 collection cups and 500 centrifuge tubes with caps for 500 determinations. The Count-10 Urine Collection System with K-Tubes includes 500 collection cups and 500 unbreakable 12mL centrifuge tubes made with resilient K-resin for 500 determinations. The Count-10 Office Pack Urine Collection System with K-Tubes comes with 300 K-tubes, 300 Sed-Pet II™ pipets, and 300 wide-scan slides for 300 determinations.
Frequently Bought Together