Order Entry
United States
ContactUsLinkComponent
Microbumin® Microalbumin Control, Quantimetrix
Microbumin® Microalbumin Control, Quantimetrix
Catalog # 82024-194
Supplier:  Quantimetrix
CAS Number:  
undefined
Microbumin® Microalbumin Control, Quantimetrix
Catalog # 82024-194
Supplier:  Quantimetrix
Supplier Number:  1250-01
CAS Number:  

Specifications

  • Description:
    Microbumin® Microalbumin Control
  • Cat. No.:
    82024-194
  • Volume:
    10× 7 ml
  • Supplier no.:
    1250-01

Specifications

About this item

With longer stability for less waste, Microbumin Microalbumin Control monitors the performance of popular strips including Clinitek® Microalbumin Reagent Strips and Micral™ test strips.

  • 3-year refrigerated shelf life from date of manufacture
  • 6-month open vial stability when stored at 2 to 8°C
  • Liquid, made with human urine
  • Includes creatinine

Microbumin is also perfect for use with turbidimetric immunoassays using Siemens Advia®, Siemens Clinitek®, Siemens DCA 2000®, Siemens Dimension®, Beckman Coulter Synchron®, Abbott Architect®, Roche Hitachi, Ortho Vitros® and nephelometry.*

Ordering information: Box includes five vials each of Level 1 and Level 2. Refer to the package insert of currently available lots for specific analyte, stability claims and instrument compatibility.