Orders LinkContactUsLinkComponent
Para 4® Controls, Streck
Para 4® Controls, Streck
Supplier:   Streck Laboratories
Para 4® Controls, Streck
Certificates
Para 4® Controls, Streck
Para 4® Controls, Streck
14 Options Available Below
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
These four-parameter hematology controls are designed for laboratories that perform a limited number of hematology tests
The assay includes Hgb, Hct, and platelet values. It is also assayed for auxiliary methods. Controls have an open-vial stability for 14 days and a closed-vial stability for 110 days. Available in glass or plastic vials.
Product Family Options
Glass Vials
Product Information
  • Size
    Availability
    Price
  • 87005-404
    Low Level
    Low Level
    1.5 mL
    Log in for availability
    Log in for availability
    Box of 6
    Box of 6 - $91.68
    $91.68
    Box of 6
       $91.68
    Minimum order quantity is 1
    Supplier Number: 215426
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • 87005-402
    Normal Level
    Normal Level
    1.5 mL
    Log in for availability
    Log in for availability
    Box of 6
    Box of 6 - $100.50
    $100.50
    Box of 6
       $100.50
    Minimum order quantity is 1
    Supplier Number: 215416
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • 87005-408
    High Level
    High Level
    1.5 mL
    Log in for availability
    Log in for availability
    Box of 6
    Box of 6 - $99.91
    $99.91
    Box of 6
       $99.91
    Minimum order quantity is 1
    Supplier Number: 215436
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • 87005-392
    1 Low, 1 Normal, and 1 High Levels
    1 Low, 1 Normal, and 1 High Levels
    1.5 mL
    Log in for availability
    Log in for availability
    Box of 3
    Box of 3 - $68.07
    $68.07
    Box of 3
       $68.07
    Minimum order quantity is 1
    Supplier Number: 215403
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • 87005-394
    2 Low, 2 Normal, and 2 High Levels
    2 Low, 2 Normal, and 2 High Levels
    1.5 mL
    Log in for availability
    Log in for availability
    Box of 6
    Box of 6 - $98.24
    $98.24
    Box of 6
       $98.24
    Minimum order quantity is 1
    Supplier Number: 215406
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • 87005-400
    4 Low, 4 Normal, and 4 High Levels
    4 Low, 4 Normal, and 4 High Levels
    1.5 mL
    Log in for availability
    Log in for availability
    Box of 12
    Box of 12 - $177.05
    $177.05
    Box of 12
       $177.05
    Supplier Number: -
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • 87005-208
    6 Low, 6 Normal, and 6 High Levels
    6 Low, 6 Normal, and 6 High Levels
    1.5 mL
    Log in for availability
    Log in for availability
    Box of 18
    Box of 18 - $271.96
    $271.96
    Box of 18
       $271.96
    Minimum order quantity is 1
    Supplier Number: 215418
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Plastic Squeeze Dropper Vials
Product Information
  • Size
    Availability
    Price
  • 87005-202
    Low Level
    Low Level
    1.5 mL
    Log in for availability
    Log in for availability
    Box of 6
    Box of 6 - $102.72
    $102.72
    Box of 6
       $102.72
    Minimum order quantity is 1
    Supplier Number: 215106
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • 87005-382
    Normal Level
    Normal Level
    1.5 mL
    Log in for availability
    Log in for availability
    Box of 6
    Box of 6 - $102.72
    $102.72
    Box of 6
       $102.72
    Minimum order quantity is 1
    Supplier Number: 215107
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • 87005-204
    High Level
    High Level
    1.5 mL
    Log in for availability
    Log in for availability
    Box of 6
    Box of 6 - $102.72
    $102.72
    Box of 6
       $102.72
    Minimum order quantity is 1
    Supplier Number: 215108
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • 87005-380
    1 Low, 1 Normal, and 1 High Levels
    1 Low, 1 Normal, and 1 High Levels
    1.5 mL
    Log in for availability
    Log in for availability
    Box of 3
    Box of 3 - $68.07
    $68.07
    Box of 3
       $68.07
    Minimum order quantity is 1
    Supplier Number: 215103
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • 87005-384
    2 Low, 2 Normal, and 2 High Levels
    2 Low, 2 Normal, and 2 High Levels
    1.5 mL
    Log in for availability
    Log in for availability
    Box of 6
    Box of 6 - $91.68
    $91.68
    Box of 6
       $91.68
    Minimum order quantity is 1
    Supplier Number: 215110
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • 87005-206
    4 Low, 4 Normal, and 4 High Levels
    4 Low, 4 Normal, and 4 High Levels
    1.5 mL
    Log in for availability
    Log in for availability
    Box of 12
    Box of 12 - $195.09
    $195.09
    Box of 12
       $195.09
    Minimum order quantity is 1
    Supplier Number: 215112
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • 87005-388
    4 Normal and 4 High Levels
    4 Normal and 4 High Levels
    1.5 mL
    Log in for availability
    Log in for availability
    Box of 8
    Box of 8 - $119.64
    $119.64
    Box of 8
       $119.64
    Minimum order quantity is 1
    Supplier Number: 215115
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.