Sweat Control® For Cyctic Fibrosis Testing, Quantimetrix
Catalog # 82024-200
Supplier: Quantimetrix
CAS Number:
Specifications
- Description:Sweat Control® For Cystic Fibrosis Testing
- Cat. No.:82024-200
- Volume:9× 3 ml
- Includes:Level 1, 2 and 3
- Supplier no.:1190-01
Specifications
About this item
Sweat Control features extended stability for convenience and less waste. Monitors the performance of cystic fibrosis assay methods including Chloridometer, Osmometer, Sweat-Chek™ conductivity analyzer and Dimension®, Advia and Hitachi ISE electrodes.
- 5-year refrigerated shelf life from date of manufacture
- 3-month open vial stability when stored at 2 to 8°C
- Liquid, simulated human sweat matrix
Ordering information: Box includes three vials each of Level 1, Level 2 and Level 3. Refer to the package insert of currently available lots for specific analyte, stability claims and instrument compatibility.