Spinalscopics® Spinal Fluid Cell Count Control, Quantimetrix
Supplier: Quantimetrix
Certificates
About this item
Use Spinalscopics Spinal Fluid Cell Count Control to monitor the performance of red and white blood cell count methods in cerebrospinal fluid samples performed manually using a hemocytometer. Easy-to-use dropper dispenser.
- Liquid, human protein matrix with stabilized human cells
- 18-month refrigerated shelf life from date of manufacture
- 6-month open vial stability when stored at 2 to 8°C
- 1-month open vial stability at room temperature
Ordering information: Refer to the package insert of currently available lots for specific analyte, stability claims and instrument compatibility.
Frequently Bought Together